ENA187622739 Enamine

2-[5-(morpholin-4-yl)-4-(prop-2-en-1-yl)-4H-1,2,4-triazol-3-yl]acetonitrile hydrobromide

  • MDL number MFCD11857872
  • Cas Number : 1171589-44-8
  • Molecular Weight : 314.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-42171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42171
Availability details for
Quantity
  • Availability
  • Add to Cart