ENA187622743 Enamine

(cyclopropylmethyl)hydrazine hydrochloride

  • MDL number MFCD11857876
  • Cas Number : 1181457-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42188
Availability details for
Quantity
  • Availability
  • Add to Cart