ENA187622756 Enamine

2-(ethylamino)-N-[4-(morpholin-4-yl)phenyl]acetamide hydrochloride

  • MDL number MFCD11857892
  • Cas Number : 1170174-36-3
  • Molecular Weight : 299.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42344
Availability details for
Quantity
  • Availability
  • Add to Cart