ENA187622767 Enamine

2-(1-tert-butyl-1H-pyrazol-4-yl)-2-oxoacetic acid

  • MDL number MFCD11857904
  • Cas Number : 1171422-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42485
Availability details for
Quantity
  • Availability
  • Add to Cart