ENA187622769 Enamine

3-(3-methoxyphenyl)-2-(5-methyl-1H-1,2,3,4-tetrazol-1-yl)propanoic acid

  • MDL number MFCD11857906
  • Cas Number : 1118787-15-7
  • Molecular Weight : 262.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-42511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42511
Availability details for
Quantity
  • Availability
  • Add to Cart