ENA187622771 Enamine

N-cyclopropyl-2-oxo-2-(piperazin-1-yl)acetamide hydrochloride

  • MDL number MFCD11857909
  • Cas Number : 1171934-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42516
Availability details for
Quantity
  • Availability
  • Add to Cart