ENA187622772 Enamine

4-(aminomethyl)-N-cyclopropylbenzene-1-sulfonamide hydrochloride

  • MDL number MFCD11857910
  • Cas Number : 1170903-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42518
Availability details for
Quantity
  • Availability
  • Add to Cart