ENA187622776 Enamine

4-amino-N-[3-(dimethylamino)propyl]benzene-1-sulfonamide dihydrochloride

  • MDL number MFCD11857913
  • Cas Number : 1171911-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42561
Availability details for
Quantity
  • Availability
  • Add to Cart