ENA187622777 Enamine

1-methyl-N-(piperidin-4-yl)-1H-pyrazole-4-carboxamide hydrochloride

  • MDL number MFCD11857914
  • Cas Number : 1173078-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 294...296
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42563

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42563
Availability details for
Quantity
  • Availability
  • Add to Cart