ENA187622778 Enamine

3-amino-1-[4-(4-fluorophenyl)piperazin-1-yl]propan-1-one dihydrochloride

  • MDL number MFCD11857916
  • Cas Number : 1172550-38-7
  • Molecular Weight : 324.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42569
Availability details for
Quantity
  • Availability
  • Add to Cart