ENA187622782 Enamine

3,5-dichloro-N-[(2-methoxyphenyl)methyl]aniline hydrochloride

  • MDL number MFCD11857921
  • Cas Number : 1171507-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167...169
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42585

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42585
Availability details for
Quantity
  • Availability
  • Add to Cart