ENA187622789 Enamine

methyl[2-phenyl-2-(propan-2-yloxy)ethyl]amine hydrochloride

  • MDL number MFCD11857931
  • Cas Number : 1172082-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42634
Availability details for
Quantity
  • Availability
  • Add to Cart