ENA187622807 Enamine

cyclopentyl(4-fluorophenyl)methanamine hydrochloride

  • MDL number MFCD11813964
  • Cas Number : 1171555-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-42045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42045
Availability details for
Quantity
  • Availability
  • Add to Cart