ENA187622811 Enamine

1-(4-fluorophenyl)-2,2-dimethylpropan-1-amine

  • MDL number MFCD09812079
  • Cas Number : 1016506-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-42050

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42050
Availability details for
Quantity
  • Availability
  • Add to Cart