ENA187622812 Enamine

2,6,6-trimethyl-4,5,6,7-tetrahydro-1-benzofuran-4-amine

  • MDL number MFCD09936557
  • Cas Number : 1016796-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42051
Availability details for
Quantity
  • Availability
  • Add to Cart