ENA187622815 Enamine

2-(3-aminophenyl)-6-ethyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD11840130
  • Cas Number : 1118787-02-2
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-42063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42063
Availability details for
Quantity
  • Availability
  • Add to Cart