ENA187622825 Enamine

1-[4-(2-methoxyethoxy)phenyl]ethan-1-one

  • MDL number MFCD09944383
  • Cas Number : 60893-75-6
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-42075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42075
Availability details for
Quantity
  • Availability
  • Add to Cart