ENA187622838 Enamine

2-(1,3-dioxaindan-5-yloxy)ethan-1-amine hydrochloride

  • MDL number MFCD11821418
  • Cas Number : 1171507-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-42095

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42095
Availability details for
Quantity
  • Availability
  • Add to Cart