ENA187622839 Enamine

5-amino-1-benzofuran-2-carboxamide hydrochloride

  • MDL number MFCD11839871
  • Cas Number : 1171762-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 300...302
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42096

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42096
Availability details for
Quantity
  • Availability
  • Add to Cart