ENA187622843 Enamine

2-(naphthalen-2-yl)oxirane

  • MDL number MFCD01712738
  • Cas Number : 20861-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-42100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42100
Availability details for
Quantity
  • Availability
  • Add to Cart