ENA187622855 Enamine

1-(1,3-dioxaindan-5-yl)cyclopropane-1-carbonitrile

  • MDL number MFCD10692862
  • Cas Number : 33522-14-4
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42114
Availability details for
Quantity
  • Availability
  • Add to Cart