ENA187622859 Enamine

4-[(piperidin-4-yl)methyl]morpholine

  • MDL number MFCD05215184
  • Cas Number : 81310-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-42119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42119
Availability details for
Quantity
  • Availability
  • Add to Cart