ENA187622865 Enamine

2,4-dioxo-1H,2H,3H,4H-pyrido[2,3-d]pyrimidine-6-carboxylic acid

  • MDL number MFCD02167459
  • Cas Number : 343347-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42131
Availability details for
Quantity
  • Availability
  • Add to Cart