ENA187622870 Enamine

5-[(naphthalen-1-yl)methyl]-1,3,4-oxadiazol-2-amine

  • MDL number MFCD02671875
  • Cas Number : 500867-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42142
Availability details for
Quantity
  • Availability
  • Add to Cart