ENA187622873 Enamine

5-[(3-fluorophenyl)methyl]-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09814180
  • Cas Number : 1016497-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42145
Availability details for
Quantity
  • Availability
  • Add to Cart