ENA187622884 Enamine

1-{4-[2-(dimethylamino)ethoxy]-3-methoxyphenyl}methanamine dihydrochloride

  • MDL number MFCD11802532
  • Cas Number : 1170882-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42161
Availability details for
Quantity
  • Availability
  • Add to Cart