ENA187622898 Enamine

dichloro-1,2-thiazole-4-carbonitrile

  • MDL number MFCD00227538
  • Cas Number : 2120-82-3
  • Molecular Weight : 179.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-66
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-42183

Safety Information

signalword Danger
pcodes P264,P270,P301+P310,P330,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-42183
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart