ENA187622906 Enamine

1-(2-aminophenyl)-3-(propan-2-yl)urea

  • MDL number MFCD11099928
  • Cas Number : 224951-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42193

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42193
Availability details for
Quantity
  • Availability
  • Add to Cart