ENA187622908 Enamine

[(piperidin-3-yl)methyl]urea

  • MDL number MFCD09810022
  • Cas Number : 1016751-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42195
Availability details for
Quantity
  • Availability
  • Add to Cart