ENA187622914 Enamine

5-phenyl-1-(pyridin-2-yl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD09808578
  • Cas Number : 80884-14-6
  • Molecular Weight : 265.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42206
Availability details for
Quantity
  • Availability
  • Add to Cart