ENA187622921 Enamine

N-(3-amino-4-methylphenyl)benzamide

  • MDL number MFCD02046983
  • Cas Number : 221875-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42213
Availability details for
Quantity
  • Availability
  • Add to Cart