ENA187622935 Enamine

3-tert-butyl-3-azabicyclo[3.2.1]octan-8-one

  • MDL number MFCD13806387
  • Cas Number : 1087789-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42232
Availability details for
Quantity
  • Availability
  • Add to Cart