ENA187622940 Enamine

3-(5,6-difluoro-1H-1,3-benzodiazol-1-yl)propanoic acid

  • MDL number MFCD09815709
  • Cas Number : 1016515-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157...159
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39713

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39713
Availability details for
Quantity
  • Availability
  • Add to Cart