ENA187622951 Enamine

2-methoxypyridin-3-amine

  • MDL number MFCD00833386
  • Cas Number : 20265-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42256

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-42256
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart