ENA187622953 Enamine

2-(2-oxoazocan-1-yl)acetic acid

  • MDL number MFCD09803625
  • Cas Number : 1016699-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42261
Availability details for
Quantity
  • Availability
  • Add to Cart