ENA187622957 Enamine

3-methyl-2-(3-methylbutanamido)butanoic acid

  • MDL number MFCD08444947
  • Cas Number : 91108-84-8
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42274
Availability details for
Quantity
  • Availability
  • Add to Cart