ENA187622965 Enamine

3-(cyclopentanesulfonyl)propanoic acid

  • MDL number MFCD09951481
  • Cas Number : 933710-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42290
Availability details for
Quantity
  • Availability
  • Add to Cart