ENA187622968 Enamine

N-(1H-1,3-benzodiazol-2-ylmethyl)furan-2-carboxamide

  • MDL number MFCD02984493
  • Cas Number : 378208-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42305
Availability details for
Quantity
  • Availability
  • Add to Cart