ENA187622980 Enamine

11-oxa-1,8-diazatricyclo[7.5.0.0,2,7]tetradeca-2,4,6,8-tetraen-13-ol

  • MDL number MFCD02578610
  • Cas Number : 68857-67-0
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-42318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42318
Availability details for
Quantity
  • Availability
  • Add to Cart