ENA187622981 Enamine

3-(3-aminophenyl)-1-methylimidazolidine-2,4-dione

  • MDL number MFCD11857888
  • Cas Number : 1114824-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42319
Availability details for
Quantity
  • Availability
  • Add to Cart