ENA187622987 Enamine

1-(2-amino-2-phenylethyl)pyrrolidin-2-one

  • MDL number MFCD09740366
  • Cas Number : 43003-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-42339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42339
Availability details for
Quantity
  • Availability
  • Add to Cart