ENA187622991 Enamine

2-hydroxy-N-(4-methylphenyl)acetamide

  • MDL number MFCD08442157
  • Cas Number : 102878-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42345
Availability details for
Quantity
  • Availability
  • Add to Cart