ENA187622993 Enamine

2-(cyclopropylamino)acetic acid

  • MDL number MFCD11185506
  • Cas Number : 10294-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210...212
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42347

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42347
Availability details for
Quantity
  • Availability
  • Add to Cart