ENA187622998 Enamine

2,6-dichloro-4-(trifluoromethyl)benzoic acid

  • MDL number MFCD01934568
  • Cas Number : 189338-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140...142
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42352

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42352
Availability details for
Quantity
  • Availability
  • Add to Cart