ENA187623002 Enamine

3-(aminomethyl)-N-(butan-2-yl)benzamide

  • MDL number MFCD09050791
  • Cas Number : 926229-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42361
Availability details for
Quantity
  • Availability
  • Add to Cart