ENA187623003 Enamine

3-carbamothioylbenzamide

  • MDL number MFCD09937067
  • Cas Number : 106748-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210...212
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40822

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40822
Availability details for
Quantity
  • Availability
  • Add to Cart