ENA187623006 Enamine

2-({[5-(trifluoromethyl)pyridin-2-yl]sulfanyl}methyl)aniline

  • MDL number MFCD11857896
  • Cas Number : 144214-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74...76
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42366

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42366
Availability details for
Quantity
  • Availability
  • Add to Cart