ENA187623007 Enamine

N1-(pyridin-2-yl)propane-1,3-diamine

  • MDL number MFCD09931684
  • Cas Number : 38585-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-42368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42368
Availability details for
Quantity
  • Availability
  • Add to Cart