ENA187623010 Enamine

3,3-dimethyl-1-(1,2,3,4-tetrahydroisoquinolin-2-yl)butan-2-amine

  • MDL number MFCD09736039
  • Cas Number : 953898-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42395
Availability details for
Quantity
  • Availability
  • Add to Cart