ENA187623018 Enamine

1-(4-aminophenyl)-N-(4-methoxyphenyl)methanesulfonamide

  • MDL number MFCD11833084
  • Cas Number : 1040033-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42403

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42403
Availability details for
Quantity
  • Availability
  • Add to Cart